
α-Bromo-2,6-dichlorotoluene
Catalog # APOSOR2114-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:239.93 g/mol
Boiling Pt:135 °C/9.75mmHg
Melting Pt:54…57 °C
Flash Pt:>112 °C
Storage Temperature:Ambient
EINECS:243-827-4
UN:3261
ADR:8,II
:GHS05
CAS Number:20443-98-5
Formula:C₇H₅BrCl₂
MDL Number:MFCD00000577