Order Entry
Spain
ContactUsLinkComponent
α-Bromo-2,3-dichlorotoluene 98%
Corrosive
α-Bromo-2,3-dichlorotoluene 98%
Catalog # APOSOR16672-25G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
α-Bromo-2,3-dichlorotoluene 98%
Catalog # APOSOR16672-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :239.93 g/mol
Boiling Pt:140…144 °C/31mmHg
Melting Pt:29…32 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :GHS05
CAS Number:57915-78-3
Formula:C₇H₅BrCl₂
MDL Number:MFCD03701317