1-Bromo-2,6-dichlorobenzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:19393-92-1
:
Formula:C₆H₃BrCl₂
MDL Number:MFCD00000574
:225.9 g/mol
Boiling Pt:241…243 °C
Melting Pt:59…64 °C
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:243-018-6
:GHS07
Frequently Bought Together