
3-Bromobenzaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3132-99-8
:
Formula:C₇H₅BrO
MDL Number:MFCD00003345
Boiling Pt:233…236 °C
Melting Pt:18…21 °C
Density:1.587 g/cm³ (25 °C)
Flash Pt:96 °C
:185.02 g/mol
EINECS:221-526-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together