Order Entry
Spain
ContactUsLinkComponent
5-Bromo-2-chloroanisole
Irritant
5-Bromo-2-chloroanisole
Catalog # APOSOR0177-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Bromo-2-chloroanisole
Catalog # APOSOR0177-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :221.48 g/mol
Boiling Pt:240…245 °C
Melting Pt:27…28 °C
Density:1.631 g/cm³ (25 °C)
Flash Pt:113 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:16817-43-9
Formula:C₇H₆BrClO
MDL Number:MFCD00078659