Tin 99.999%, powder –100 mesh
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7440-31-5
:
Formula:Sn
MDL Number:MFCD00133862
Boiling Pt:2270 °C
Melting Pt:231.9 °C
Density:7.29 g/cm³ (25 °C)
Molecular Weight:118.71 g/mol
Merck Index:12,09587
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together