

Palladium(II) acetate 99.95+%
Catalog # APOSIN3435-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:224.51 g/mol
Melting Pt:216.3…223.7 °C (dec.)
EINECS:222-164-4
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:3375-31-3
Formula:C₄H₆O₄Pd
MDL Number:MFCD00012453