Order Entry
Spain
Orders LinkContactUsLinkComponent
Silver nitrate 99.9+%

Silver nitrate 99.9+%

Oxidizing
Corrosive
Supplier:   Apollo Scientific
Silver nitrate 99.9+%
Supplier:  Apollo Scientific
Certificates
Silver nitrate 99.9+%

2 Options Available Below

Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Melting Pt:212 °C (dec.)
  • Density:4.352 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:231-853-9
  • UN:1493
  • ADR:5.1,II

Documents

Product Family Options

Product Information