Order Entry
Spain
ContactUsLinkComponent
SKF-86002
SKF-86002
Catalog # APOSBISN0228-10MG
Supplier:  Apollo Scientific
CAS Number:  
SKF-86002
Catalog # APOSBISN0228-10MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :297.35 g/mol
Storage Temperature:Ambient
CAS Number:72873-74-6
Formula:C₁₆H₁₂F₃FS
MDL Number:MFCD00869367