
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7240-90-6
:
Formula:C₁₄H₁₅BrClNO₆
MDL Number:MFCD00005666
:408.63 g/mol
Melting Pt:230 °C
EINECS:230-640-8
:GHS07
Storage Temperature:Freezer
Frequently Bought Together