γ-Cyclodextrine
Bottle, 50 g
Catalog # APOSBIM1016-50G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
- CAS Number:17465-86-0
- Formula:C₄₈H₈₀O₄₀
- MDL Number:MFCD00009595
- Molecular Weight:1297.1248 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



