L-Ergothioneine
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: (S)-3-(2-Thioxo-2,3-dihydro-1H-imidazol-4-yl)-2-(trimethylammonio)propanoate, Ergothioneine
CAS Number:497-30-3
:
Formula:C₉H₁₅N₃O₂S
MDL Number:MFCD00167474
Molecular Weight:229.3 g/mol
Melting Pt:255…259 °C
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together