L(+)-Glutamic acid ≥99%
Bottle, 500 g
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Ph.Eur, Pyrogen free
Synonyms: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid
- CAS Number:56-86-0
- Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
- MDL Number:MFCD00002634
- Molecular Weight:147.13 g/mol
- Melting Pt:205 °C (dec.)
- Density:1.525 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:200-293-7
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Pk:500 g



