L(-)-Glutathione (reduced form)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:70-18-8
:
Formula:C₁₀H₁₇N₃O₆S
MDL Number:MFCD00065939
:307.33 g/mol
Melting Pt:192…195 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together