Order Entry
Spain
ContactUsLinkComponent
N-t-BOC-cis-4-Fluoro-L-proline methyl ester 95+%
N-t-BOC-cis-4-Fluoro-L-proline methyl ester 95+%
Catalog # APOSPC409006-1G
Supplier:  Apollo Scientific
CAS Number:  
N-t-BOC-cis-4-Fluoro-L-proline methyl ester 95+%
Catalog # APOSPC409006-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :247.263 g/mol
Storage Temperature:Ambient
CAS Number:203866-16-4
Formula:C₁₁H₁₈FNO₄
MDL Number:MFCD11054130