Order Entry
Spain
ContactUsLinkComponent
1,5-Dibromo-2,3-dichlorobenzene 97%
1,5-Dibromo-2,3-dichlorobenzene 97%
Catalog # APOSOR71002-1G
Supplier:  Apollo Scientific
CAS Number:  
1,5-Dibromo-2,3-dichlorobenzene 97%
Catalog # APOSOR71002-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :304.794 g/mol
Storage Temperature:Refrigerator
CAS Number:81067-42-7
Formula:C₆H₂Br₂Cl₂
MDL Number:MFCD00070783