Order Entry
Spain
ContactUsLinkComponent
2,4-Dichlorobenzene-1,3-diol 95%
2,4-Dichlorobenzene-1,3-diol 95%
Catalog # APOSOR480689-25G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Dichlorobenzene-1,3-diol 95%
Catalog # APOSOR480689-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :179.001 g/mol
Storage Temperature:Ambient
CAS Number:16606-61-4
Formula:C₆H₄Cl₂O₂
MDL Number:MFCD13152136