N'-[(1E)-1-(4-Bromophenyl)ethylidene]methoxycarbohydrazide, tech
Catalog # APOSOR32068-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:271.111 g/mol
Storage Temperature:Ambient
CAS Number:1211461-40-3
Formula:C₁₀H₁₁BrN₂O₂
MDL Number:MFCD00792940