
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1214264-88-6
:
Formula:C₁₆H₂₀B₂N₂O₂
MDL Number:MFCD27980593
:293.964 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together