Erioglaucine disodium salt, Sigma-Aldrich®
:3844-45-9
:
:C₃₇H₃₄Na₂N₂O₉S₃
:MFCD00012141
:792.84 g/mol
:283 °C (dec., lit.)
:M-Clarity™ quality level = MQ200
:Room Temperature
- Appearance (Colour):Purple to Dark Purple
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Wavelength
c = 0.005 g/l; Water:627 - 631 nm - Extinction Coefficient:≥80000
- Wavelength:405 - 409 nm
- Thin Layer Chromatography:Consistent with Past Lots
- Solubility (Turbidity)
c = 1 mg/ml; Water:Clear - Solubility (Colour):Dark Blue
- Carbon:39.2 - 72.9 %
- Nitrogen:2.4 - 4.6 %
- Dye Content:Based on Nitrogen Content
Frequently Bought Together