Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Disperse Yellow 3
Disperse Yellow 3 30% (dye content), Sigma-Aldrich®
Disperse Yellow 3 30% (dye content), Sigma-Aldrich®
215686-50G
:
SIGMA ALDRICH MICROSCOPY
:
Disperse Yellow 3 30% (dye content), Sigma-Aldrich®
215686-50G
:
SIGMA ALDRICH MICROSCOPY
:
Pk:
50 g
Seq:
3368
:
:
269.3 g/mol
:
268…270 °C (lit.)
:
GHS08
:
GHS07
:
Ambient
:
2832-40-8
:
C₁₅H₁₅N₃O₂
:
MFCD00002377