Order Entry
Spain
ContactUsLinkComponent
Leukocyte Alkaline Phosphatase Kit based on naphthol AS-MX phosphate and fast blue RR salt (with citrate), Sigma-Aldrich®
Corrosive
Leukocyte Alkaline Phosphatase Kit based on naphthol AS-MX phosphate and fast blue RR salt (with citrate), Sigma-Aldrich®
Catalog # 85L2-1KT
CAS Number:  
Corrosive
Leukocyte Alkaline Phosphatase Kit based on naphthol AS-MX phosphate and fast blue RR salt (with citrate), Sigma-Aldrich®
Catalog # 85L2-1KT
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ600 (more info.)
 :GHS05
Storage Temperature:Refrigerator
 :1