
Simethicone
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:8050-81-5
:
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to grey Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4000 to 1.4100 (20°C, 589 nm)
- Viscosity:475 to 525 CP (25°C)
Specification Test Results
Frequently Bought Together