Order Entry
Spain
ContactUsLinkComponent
VWR® Aluminium Foil Seals for PCR
VWR® Aluminium Foil Seals for PCR
Catalog # 391-0641
Supplier:  VWR Collection
undefined
VWR® Aluminium Foil Seals for PCR
Catalog # 391-0641
Supplier:  VWR Collection

Specifications

  • Optical property:
    Light blocking
  • Sterile:
    -
  • Description:
    Sealing foil, roll format
  • Thickness:
    87,6 µm
  • Pk:
    500

Specifications

About this item

Pre-cut aluminum foil seals on a roll for PCR applications. Supplied on a roll with holder for ease of use.

  • Ideal for PCR
  • Rapidly seal microplates; faster than applying single sheeted film
  • 2 mil pierceable foil
  • Certified DNase- and RNase-free
  • Recommended temperature range from −20 to +120 °C

Self-standing, non slip dispenser is supplied for ease of use.