exsig® COVID-19 direct
Catalog # ZEXSIGCOV19DIR
Supplier: PRIMERDESIGN
CAS Number:
Specifications
- Pk:96 Tests
- Description:Coronavirus (COVID-19) Real-Time qPCR assay
Specifications
About this item
A total workflow solution with all reagents and components provided from start to finish.
- For use with dry swabs for a rapid and sensitive workflow
- Validated against CFX (Bio-Rad), ABI 7500 (Applied Biosystems) and LC480 II (Roche)
- Kit size 96 (preparations and reactions)
- Master mix included
- Kit targets the ORF1ab region of SARS-CoV-2, and has 100% homology with all publicly available sequences
Certifications: CE-IVD
Accessories information: Traceability of values assigned to calibrating of control materials. Assured through reference measurement proceduresComprehensive validation in concordance with ISO13485. Post-market maintenance of technical files, including strain information. Total kit performance measurement and assurance provided.
Delivery: Lyophilised kits with no dry ice shipping requirement.
RNase inhibitor recommended (not included).
RNase inhibitor recommended (not included).