Order Entry
Spain
ContactUsLinkComponent
Cyanide standard solution, traceable to SRM from NIST, Certipur®, Supelco®
Cyanide standard solution, traceable to SRM from NIST, Certipur®, Supelco®
Catalog # 1.19533.0500
Supplier:  Merck
CAS Number:  
Cyanide standard solution, traceable to SRM from NIST, Certipur®, Supelco®
Catalog # 1.19533.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Cyanide standard solution
  • M-Clarity quality level:
    MQ100
  • Contents:
    K₂[Zn(CN)₄] in H₂O

Specifications

About this item