Order Entry
Spain
Orders LinkContactUsLinkComponent
Clamps, universal
Clamps, universal
0 - 80 mm
Catalog # 241-7192
Supplier:   BOCHEM
Clamps, universal
Catalog # 241-7192
Supplier:  BOCHEM
Clamps, universal
Clamps, universal
0 - 80 mm
Price
23,60 €
Log in for availability
1 * 1 UN
Product Details & Documents
Aluminium.
  • The fingers of the clamps have cork inserts to protect the clamped items
  • Without bosshead for fixing to stand
  • With wing screws for rapid easy clamping
Specifications
  • Spanning width:
    0 - 80 mm