Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Ficin
Ficin
Ficin
Catalog # TCIAF0010-25G
Supplier:
TCI
CAS Number:
Expand image
Ficin
Catalog # TCIAF0010-25G
Supplier:
TCI
CAS Number:
Specifications
Pk:
25 g
Enzyme name:
Ficin
Specifications
About this item
Product Details
Product Details