Ammonium metavanadate ≥99.0% analytical reagent
21369.186
:
- Pack type:Plastic bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
- Assay:Min. 99.0 %
- Cl (Chloride):Max. 50 ppm
- SO4 (Sulphate):Max. 0.20 %
- Pb (Lead):Max. 20 ppm
: Ammonium monovanadate, Ammonium vanadium trioxide, Ammonium vanadium oxide
:
:116.98 g/mol
:232-261-3
:2859
:GHS06
:7803-55-6
:NH₄VO₃
:MFCD00011430
:200
:2.33 g/cm³ (20 °C)
:6.1 II
:13,00572