



Calcium hypochlorite for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7778-54-3
:
Formula:CaCl₂O₂
MDL Number:MFCD00010900
:142.98 g/mol
Merck Index:13,02111
Melting Pt:∼100 °C
Density:2.35 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-908-7
ADR:5.1,II
:M-Clarity™ quality level = MQ200
UN:1748
:GHS03
:GHS05
:GHS07
:GHS09
Specification Test Results
- Assay (iodometric):67.0 - 75.0 %
- Identity:passes test
- Molar Mass:142.98 g/mol
- Density:2.35 g/cm3 (20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
Frequently Bought Together