

Isophorone 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-59-1
:
Formula:C₉H₁₄O
MDL Number:MFCD00001584
:138.21 g/mol
Merck Index:13,05215
Boiling Pt:213…214 °C (760 mmHg)
Melting Pt:–8 °C
Density:0.92 g/cm³
Flash Pt:84 °C
Storage Temperature:Ambient
EINECS:201-126-0
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4750 to 1.4800 (20°C, 589 nm)
- GC:≥96.0 %
- NMR:consistent with structure
Specification Test Results
Frequently Bought Together