Xylene fast yellow 2G
TCIAX0025-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:Light yellow to Brown powder to crystal
- Lambda max.:401.0 to 405.0 nm (H2O)
- Absorbance (E1%1cm):min. 350 (H2O, 401.0 to 405.0 nm)
:
:551.28 g/mol
:228-819-0
:6359-98-4
:C₁₆H₁₂Cl₂N₄O₇S₂
:MFCD00053367