Wedelolactone ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:524-12-9
:
Formula:C₁₆H₁₀O₇
MDL Number:MFCD07778564
:314.2 g/mol
Melting Pt:315°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
Frequently Bought Together