Order Entry
Spain
ContactUsLinkComponent
 
4,4',4''-(1,3,5-Triazine-2,4,6-triyl)trianiline ≥98.0% (by HPLC)
 :  TCI
 
4,4',4''-(1,3,5-Triazine-2,4,6-triyl)trianiline ≥98.0% (by HPLC)
 :  TCI

 

  • Appearance:
    White to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 95.0 %

 

Frequently Bought Together