4-(Trifluoromethyl)benzamide ≥98.0% (by GC)
: TCI
:1891-90-3
:
:C₈H₆F₃NO
:MFCD00007998
:189.14 g/mol
:217-571-9
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:183.0 to 187.0 deg-C
Frequently Bought Together