Order Entry
Spain
ContactUsLinkComponent
DL-Threonine ≥97.0% (by titrimetric analysis) (contains DL-Allothreonine)
DL-Threonine ≥97.0% (by titrimetric analysis) (contains DL-Allothreonine)
Supplier:  TCI
Certificates
undefined
DL-Threonine ≥97.0% (by titrimetric analysis) (contains DL-Allothreonine)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 97.0 %

Specification Test Results

Frequently Bought Together