Nε-trifluoroacetyl-L-lysine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:10009-20-8
:
Formula:C₈H₁₃F₃N₂O₃
MDL Number:MFCD00037223
:242.20 g/mol
Melting Pt:254°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:+17.0 to +20.0 deg (C=1, 1mol/L HCl)
Specification Test Results
Frequently Bought Together