Order Entry
Spain
ContactUsLinkComponent
 
L-Tyrosine-tert-butyl ester ≥97.0% (by HPLC, titration analysis)
 :  TCI
 
L-Tyrosine-tert-butyl ester ≥97.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    140.0 to 145.0 deg-C
  • Specific rotation [a]20/D:
    +23.0 to +27.0 deg (C=2, EtOH)

 

Frequently Bought Together