Tetrakis(trifluoro-2,4-pentanedionato)zirconium(IV) ≥98.0% (by titrimetric analysis)
Catalog # TCIAT0747-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
- Melting point:129.0 to 133.0 deg-C
Specification Test Results
About this item
:
:703.54 g/mol
CAS Number:17499-68-2
Formula:C₂₀H₁₆F₁₂O₈Zr
MDL Number:MFCD00015317