Order Entry
Spain
ContactUsLinkComponent
Hanks’ balanced salt solutions (HBSS) modified 10x without calcium, magnesium or sodium bicarbonate, Sigma-Aldrich®
Hanks’ balanced salt solutions (HBSS) modified 10x without calcium, magnesium or sodium bicarbonate, Sigma-Aldrich®
Certificates
Hanks’ balanced salt solutions (HBSS) modified 10x without calcium, magnesium or sodium bicarbonate, Sigma-Aldrich®

Specification Test Results

  • Appearance (Colour):
    Reddish-Orange
  • Appearance (Form):
    Solution
  • Appearance (Turbidity)
    Essentially free of particles:
    Clear
  • pH
    At 25 degrees C.:
    5.8 - 6.0
  • Osmolality:
    245.0 - 275.0 mOs/kg
  • Microbiological Test:
    Pass

Specification Test Results

Related Information

Frequently Bought Together