DL-Sodium lactate 60% in water
Catalog # TCIAS0928-500G
Supplier: TCI
CAS Number:
Specifications
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Nonaqueous Titration):60.0 to 65.0 w/w%
Specification Test Results
About this item
:
:112.06 g/mol
Merck Index:14,08635
EINECS:200-772-0
:GHS07
CAS Number:72-17-3
:312-85-6
Formula:H₃CCH(OH)COONa
MDL Number:MFCD00065400