Order Entry
Spain
ContactUsLinkComponent
Sunset yellow FCF ≥90.0% (by UV/Vis), crystals
Sunset yellow FCF ≥90.0% (by UV/Vis), crystals
Catalog # TCIAS0141-25G
Supplier:  TCI
CAS Number:  
undefined
Sunset yellow FCF ≥90.0% (by UV/Vis), crystals
Catalog # TCIAS0141-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (UV-vis method):
    min. 90.0 % (calcd.on dried substance)
  • Drying loss:
    max. 10.0 %

Specification Test Results

About this item

 :
 :452.36 g/mol
Merck Index:14,09001
EINECS:220-491-7
CAS Number:2783-94-0
Formula:C₁₆H₁₀N₂Na₂O₇S₂
MDL Number:MFCD00036437