Order Entry
Spain
ContactUsLinkComponent
 
Palladium 5% on calcium carbonate for use as a catalyst lead poisoned
 :  TCI
 
Palladium 5% on calcium carbonate for use as a catalyst lead poisoned
 :  TCI

 

  • Appearance:
    Light gray to Brown to Black powder to crystal
  • Content (Palladium):
    4.90 to 5.10 %
  • Lead:
    2.40 to 3.00 %

 

Frequently Bought Together