
N-(1-Pyrenyl)maleimide ≥97.0% (by HPLC)
Catalog # TCIAP1214-250MG
Supplier: TCI
CAS Number:
Specifications
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 97.0 area%
Specification Test Results
About this item
:
:297.31 g/mol
EINECS:255-702-1
:GHS07
CAS Number:42189-56-0
Formula:C₂₀H₁₁NO₂
MDL Number:MFCD00049301