Protamine sulphate (from salmon sperm)
Supplier: TCI
Certificates
About this item
CAS Number:53597-25-4
:9009-65-8
MDL Number:MFCD00132091
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Total Nitrogen:21.0 to 26.0 % (after drying)
- Specific rotation [a]20/D:-68.0 to -72.0 deg (C=2, H2O)
- Solubility in hot Water:almost transparency
- Drying loss:max. 10.0 %
Specification Test Results
Frequently Bought Together