Order Entry
Spain
ContactUsLinkComponent
 
 
2-Phenylethylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
 
2-Phenylethylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Solubility in Water:
    almost transparency

 

 

Frequently Bought Together