
Ethyl cinnamate for synthesis, Sigma-Aldrich®
Catalog # 8.00238.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 ml
- Seq:2590
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:6 - 7 °C
- Molar Mass:176.21 g/mol
- Boiling Point:271 °C (1013 hPa)
- Flash Point:135 °C
- Refractive Index:1.5585 (20 °C, 589 nm)
- Density:1.05 g/cm3 (20 °C)
- Synonyms:Ethyl trans-3-phenylpropenoate, Cinnamic acid ethyl ester
- Hill Formula:C11H12O2
- Chemical Formula:C6H5CH=CHCOOC2H5
Specification Test Results
About this item
:
:176.21 g/mol
Boiling Pt:271 °C (1013 hPa)
Melting Pt:7 °C
Density:1.046 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:203-104-6
:GHS07
CAS Number:103-36-6
Formula:C₁₁H₁₂O₂
MDL Number:MFCD00009189