Oxonol yellow K
Catalog # TCIAO0284-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Lambda max.:419.0 to 423.0 nm (H2O)
- Absorbance (E1%1cm):min. 370 (H2O, 419.0 to 423.0 nm)
Specification Test Results
About this item
:
:594.70 g/mol
CAS Number:82407-55-4
Formula:C₂₁H₁₈N₄O₈S₂
MDL Number:MFCD02093486