Picamilon ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAN0998-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:211.0 to 215.0 deg-C
Specification Test Results
About this item
:
:208.22 g/mol
CAS Number:34562-97-5
Formula:C₁₀H₁₂N₂O₃
MDL Number:MFCD00186518