Order Entry
Spain
ContactUsLinkComponent
2-(1-Naphthyl)ethanesulphonyl chloride ≥97.0% (by HPLC, titration analysis)
Corrosive
2-(1-Naphthyl)ethanesulphonyl chloride ≥97.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
2-(1-Naphthyl)ethanesulphonyl chloride ≥97.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Argentmetric Titration):
    min. 97.0 %
  • Melting point:
    46.0 to 50.0 deg-C

Specification Test Results

Frequently Bought Together